2-o-Tolyloxyethylamine 95%
Supplier: Apollo Scientific
Synonyms:
2-(2-Methylphenoxy)ethylamine
OR951264-1G
OR951264-5G
APOSOR951264-1GEA
76.1
GBP
APOSOR951264-1G
APOSOR951264-5G
2-o-Tolyloxyethylamine 95%
2-o-Tolyloxyethylamine
Formula:
C₉H₁₃NO MW: 151,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02598962 CAS Number: 26583-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...