6-tert-Butyl-m-cresol 95%
Supplier: Apollo Scientific
Synonyms:
2-tert-Butyl-5-methylphenol, 5-Methyl-2-tert-butylphenol
OR950844-25G
OR950844-100G
OR950844-500G
APOSOR950844-25GEA
48.6
GBP
APOSOR950844-25G
APOSOR950844-100G
APOSOR950844-500G
6-tert-Butyl-m-cresol 95%
6-tert-Butyl-m-cresol
Formula:
C₁₁H₁₆O MW: 164,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002308 CAS Number: 88-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...