6,6-Dimethyl-3-azabicyclo[3.1.0]hexane ≥98%
Supplier: Apollo Scientific
OR948657-25G
OR948657-5G
OR948657-10G
APOSOR948657-25GEA
178
GBP
APOSOR948657-25G
APOSOR948657-5G
APOSOR948657-10G
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane ≥98%
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane
Formula:
C₇H₁₃N Storage Temperature: Ambient |
MDL Number:
MFCD13176114 CAS Number: 943516-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...