2-Nitroimidazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Azomycin, 2-Nitro-1H-imidazole
OR9460-5G
OR9460-1G
APOSOR9460-5GEA
55.1
GBP
APOSOR9460-5G
APOSOR9460-1G
2-Nitroimidazole 97%
2-Nitroimidazole
Formula:
C₃H₃N₃O₂ MW: 113,08 g/mol Melting Pt: 287 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005185 CAS Number: 527-73-1 EINECS: 208-425-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...