4-Morpholinethiocarboxamide 95%

Supplier: Apollo Scientific

Synonyms: Morpholine-4-carbothioamide

OR944576-1G OR944576-5G
APOSOR944576-1GEA 139 GBP
APOSOR944576-1G APOSOR944576-5G
4-Morpholinethiocarboxamide 95%
4-Morpholinethiocarboxamide
Formula: C₅H₁₀N₂OS
MW: 146,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00178434
CAS Number: 14294-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR