(4-Methyl-1,2,5-oxadiazol-3-yl)methanamine

Supplier: Apollo Scientific

Synonyms: C-(4-Methylfurazan-3-yl)methylamine

OR943469-1G OR943469-250MG
APOSOR943469-1GEA 289 GBP
APOSOR943469-1G APOSOR943469-250MG
(4-Methyl-1,2,5-oxadiazol-3-yl)methanamine
(4-Methyl-1,2,5-oxadiazol-3-yl)methanamine
Formula: C₄H₇N₃O
MW: 113,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07800291
CAS Number: 321392-83-0
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR