2-Methyl-5-propionylfuran 95%
Supplier: Apollo Scientific
Synonyms:
1-(5-Methylfuran-2-yl)propan-1-one
OR942057-1G
OR942057-25G
OR942057-5G
APOSOR942057-1GEA
44.6
GBP
APOSOR942057-1G
APOSOR942057-25G
APOSOR942057-5G
2-Methyl-5-propionylfuran 95%
2-Methyl-5-propionylfuran
Formula:
C₈H₁₀O₂ MW: 138,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00053142 CAS Number: 10599-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...