4-(Isopropylamino)butanol 95%

Supplier: Apollo Scientific

OR939932-1G OR939932-5G
APOSOR939932-1GEA 178 GBP
APOSOR939932-1G APOSOR939932-5G
4-(Isopropylamino)butanol 95%
4-(Isopropylamino)butanol
Formula: C₇H₁₇NO
MW: 131,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14708173
CAS Number: 42042-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR