2-Ethylbenzonitrile 95%

Supplier: Apollo Scientific

OR939688-10G OR939688-1G OR939688-5G
APOSOR939688-10GEA 440 GBP
APOSOR939688-10G APOSOR939688-1G APOSOR939688-5G
2-Ethylbenzonitrile 95%
2-Ethylbenzonitrile
Formula: C₉H₉N
MW: 131,174 g/mol
Boiling Pt: 94…95 °C/12mmHg
Melting Pt: 51,75 °C
Density: 0,974 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00045597
CAS Number: 34136-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR