1-(1-Cyclopenten-1-yl)pyrrolidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Pyrrolidino-1-cyclopentene

OR939391-25G OR939391-5G
APOSOR939391-25GEA 565 GBP
APOSOR939391-25G APOSOR939391-5G
1-(1-Cyclopenten-1-yl)pyrrolidine 97%
1-(1-Cyclopenten-1-yl)pyrrolidine
Formula: C₉H₁₅N
MW: 137,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003162
CAS Number: 7148-07-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR