Allyl methyl carbonate 98%

Supplier: Apollo Scientific

OR939355-25G OR939355-5G OR939355-100G
APOSOR939355-25GEA 171 GBP
APOSOR939355-25G APOSOR939355-5G APOSOR939355-100G
Allyl methyl carbonate 98%
Allyl methyl carbonate
Formula: C₅H₈O₃
MW: 116,1152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134545
CAS Number: 35466-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR