1-Benzyl-1H-pyrazol-4-amine 95%
Supplier: Apollo Scientific
OR939289-1G
OR939289-250MG
APOSOR939289-1GEA
165
GBP
APOSOR939289-1G
APOSOR939289-250MG
1-Benzyl-1H-pyrazol-4-amine 95%
1-Benzyl-1H-pyrazol-4-amine
Formula:
C₁₀H₁₁N₃ Storage Temperature: Ambient |
MDL Number:
MFCD01693690 CAS Number: 28466-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...