2-Bromo-N-methylacetamide 98%
Supplier: Apollo Scientific
OR939130-10G
OR939130-1G
OR939130-5G
APOSOR939130-10GEA
780
GBP
APOSOR939130-10G
APOSOR939130-1G
APOSOR939130-5G
2-Bromo-N-methylacetamide 98%
2-Bromo-N-methylacetamide
Formula:
C₃H₆BrNO Storage Temperature: Ambient |
MDL Number:
MFCD06800288 CAS Number: 34680-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...