1-Butyl-3-methylpyridinium chloride 95%

Supplier: Apollo Scientific

Synonyms: 1-Butyl-3-methylpyridinium Chloride

OR938737-1G OR938737-5G
APOSOR938737-1GEA 48.6 GBP
APOSOR938737-1G APOSOR938737-5G
1-Butyl-3-methylpyridinium chloride 95%
1-Butyl-3-methylpyridinium chloride
Formula: C₁₀H₁₆ClN
MW: 185,7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458942
CAS Number: 125652-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR