(R)-1-Benzyl-3-pyrrolidinecarbonitrile 95%

Supplier: Apollo Scientific

Synonyms: (R)-1-Benzyl-3-cyanopyrrolidine

OR938474-1G OR938474-250MG
APOSOR938474-1GEA 565 GBP
APOSOR938474-1G APOSOR938474-250MG
(R)-1-Benzyl-3-pyrrolidinecarbonitrile 95%
(R)-1-Benzyl-3-pyrrolidinecarbonitrile
Formula: C₁₂H₁₄N₂
MW: 186,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09951967
CAS Number: 157528-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR