2-Methyl-2-(4-methylphenyl)morpholine 95%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-2-(p-tolyl)morpholine

OR938185-1G OR938185-250MG
APOSOR938185-1GEA 715 GBP
APOSOR938185-1G APOSOR938185-250MG
2-Methyl-2-(4-methylphenyl)morpholine 95%
2-Methyl-2-(4-methylphenyl)morpholine
Formula: C₁₂H₁₇NO
MW: 191,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060961
CAS Number: 902836-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR