Phenylmethanesulphonamide 98%

Supplier: Apollo Scientific

Synonyms: Alpha-toluenesulphonamide

OR938016-10G OR938016-25G OR938016-5G
APOSOR938016-10GEA 243 GBP
APOSOR938016-10G APOSOR938016-25G APOSOR938016-5G
Phenylmethanesulphonamide 98%
Phenylmethanesulphonamide
Formula: C7H9NO2S
MW: 171,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041893
CAS Number: 4563-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR