N-Allylaniline 95%

Supplier: Apollo Scientific

OR937892-25G
APOSOR937892-25GEA 120 GBP
APOSOR937892-25G
N-Allylaniline 95%
N-Allylaniline
Formula: C₉H₁₁N
MW: 133,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008638
CAS Number: 589-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR