2,5-Dimethyl-3-furanthiol 95%

Supplier: Apollo Scientific

Synonyms: 3-Mercapto-2,5-dimethylfuran, 2,5-Dimethylfuran-3-thiol

OR937853-1G OR937853-250MG
APOSOR937853-1GEA 368 GBP
APOSOR937853-1G APOSOR937853-250MG
2,5-Dimethyl-3-furanthiol 95%
2,5-Dimethyl-3-furanthiol
Formula: C₆H₈OS
MW: 128,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00801026
CAS Number: 55764-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR