1-Butylpyridinium chloride 98%
Supplier: Apollo Scientific
OR937650-25G
OR937650-5G
APOSOR937650-25GEA
0
GBP
APOSOR937650-25G
APOSOR937650-5G
1-Butylpyridinium chloride 98%
1-Butylpyridinium chloride
Formula:
C₉H₁₄ClN Storage Temperature: Ambient |
MDL Number:
MFCD00060047 CAS Number: 1124-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...