Methyl acetylacrylate 98%
Supplier: Apollo Scientific
Synonyms:
Acetylacrylic Acid Methyl Ester
OR937647-1G
OR937647-250MG
APOSOR937647-1GEA
328
GBP
APOSOR937647-1G
APOSOR937647-250MG
Methyl acetylacrylate 98%
Methyl acetylacrylate
Formula:
C₆H₈O₃ MW: 128.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059331 CAS Number: 4188-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...