(±)-1,2-Hexanediol 96%

Supplier: Apollo Scientific

Synonyms: Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane

OR937322-25G OR937322-100G
APOSOR937322-25GEA 47.2 GBP
APOSOR937322-25G APOSOR937322-100G
(±)-1,2-Hexanediol 96%
(±)-1,2-Hexanediol
Formula: C₆H₁₄O₂
MW: 118,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010737
CAS Number: 6920-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR