2-Chloro-6-methylbenzonitrile 95%

Supplier: Apollo Scientific

OR937301-1G OR937301-5G
APOSOR937301-1GEA 119 GBP
APOSOR937301-1G APOSOR937301-5G
2-Chloro-6-methylbenzonitrile 95%
2-Chloro-6-methylbenzonitrile
Formula: C₈H₆ClN
MW: 151,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001782
CAS Number: 6575-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR