Ethyl-N-methylcarbamate 95%

Supplier: Apollo Scientific

Synonyms: N-Methylurethane, N-Methylcarbamic Acid Ethyl Ester, Ethyl methylcarbamate

OR937161-25G OR937161-5G
APOSOR937161-25GEA 174 GBP
APOSOR937161-25G APOSOR937161-5G
Ethyl-N-methylcarbamate 95%
Ethyl-N-methylcarbamate
Formula: C₄H₉NO₂
MW: 103,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041924
CAS Number: 105-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR