2,2,4-Trimethyl-2-pentanol 95%

Supplier: Apollo Scientific

OR936859-1G OR936859-5G
APOSOR936859-1GEA 205 GBP
APOSOR936859-1G APOSOR936859-5G
2,2,4-Trimethyl-2-pentanol 95%
2,2,4-Trimethyl-2-pentanol
Formula: C₈H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00101611
CAS Number: 690-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR