1-Boc-pyrrole-2-carboxaldehyde 95%

Supplier: Apollo Scientific

Synonyms: tert-Butyl 2-formyl-1H-pyrrole-1-carboxylate

OR936830-1G OR936830-25G OR936830-5G
APOSOR936830-1GEA 98.3 GBP
APOSOR936830-1G APOSOR936830-25G APOSOR936830-5G
1-Boc-pyrrole-2-carboxaldehyde 95%
1-Boc-pyrrole-2-carboxaldehyde
Formula: C₁₀H₁₃NO₃
MW: 195,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03086220
CAS Number: 161282-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR