4-Butoxyphenol 95%

Supplier: Apollo Scientific

Synonyms: Hydroquinone monobutyl ether

OR936384-25G OR936384-5G
APOSOR936384-25GEA 51.2 GBP
APOSOR936384-25G APOSOR936384-5G
4-Butoxyphenol 95%
4-Butoxyphenol
Formula: C₁₀H₁₄O₂
MW: 166,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002336
CAS Number: 122-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR