Phenylsuccinic anhydride 95%

Supplier: Apollo Scientific

Synonyms: 3-Phenyldihydrofuran-2,5-dione

OR936348-25G OR936348-5G
APOSOR936348-25GEA 394 GBP
APOSOR936348-25G APOSOR936348-5G
Phenylsuccinic anhydride 95%
Phenylsuccinic anhydride
Formula: C₁₀H₈O₃
MW: 176,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192395
CAS Number: 1131-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR