4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone 98%
Supplier: Apollo Scientific
Synonyms:
4-Benzoyl-5-methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one
OR936339-10G
OR936339-50G
APOSOR936339-10GEA
114
GBP
APOSOR936339-10G
APOSOR936339-50G
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone 98%
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone
Formula:
C₁₇H₁₄N₂O₂ MW: 278,31 g/mol Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003136 CAS Number: 4551-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...