N-Ethyl-p-toluenesulphonamide 95%

Supplier: Apollo Scientific

Synonyms: N-Ethyl-4-methylbenzenesulphonamide, p-Tolueneethylsulphonamide, N-Ethyltoluene-4-sulphonamide, N-Tosylethylamine

OR936234-100G OR936234-500G
APOSOR936234-100GEA 55.1 GBP
APOSOR936234-100G APOSOR936234-500G
N-Ethyl-p-toluenesulphonamide 95%
N-Ethyl-p-toluenesulphonamide
Formula: C₉H₁₃NO₂S
MW: 199,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048511
CAS Number: 80-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR