6-Methyl-2-(2-pyridinyl)-4-pyrimidinol

Supplier: Apollo Scientific
Warning

OR9333-250MG OR9333-1G OR9333-5G
APOSOR9333-250MGEA 72.2 GBP
APOSOR9333-250MG APOSOR9333-1G APOSOR9333-5G
6-Methyl-2-(2-pyridinyl)-4-pyrimidinol
6-Methyl-2-(2-pyridinyl)-4-pyrimidinol
Formula: C₁₀H₉N₃O
MW: 187,2 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00114146
CAS Number: 55417-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR