1H-Benzotriazole-1-carboxaldehyde 95%

Supplier: Apollo Scientific

Synonyms: 1-Formyl-1H-benzotriazole, 1H-Benzo[d][1,2,3]triazole-1-carbaldehyde

OR933173-1G OR933173-5G
APOSOR933173-1GEA 263 GBP
APOSOR933173-1G APOSOR933173-5G
1H-Benzotriazole-1-carboxaldehyde 95%
1H-Benzotriazole-1-carboxaldehyde
Formula: C₇H₅N₃O
MW: 147,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00239420
CAS Number: 72773-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR