Isobutyl isovalerate 97%

Supplier: Apollo Scientific

Synonyms: Isobutyl Isovalerate, Isovaleric acid Isobutyl ester

OR932977-100G OR932977-500G
APOSOR932977-100GEA 132 GBP
APOSOR932977-100G APOSOR932977-500G
Isobutyl isovalerate 97%
Isobutyl isovalerate
Formula: C₉H₁₈O₂
MW: 158,2380 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048349
CAS Number: 589-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR