3-Chromanecarboxylic acid 95%

Supplier: Apollo Scientific

Synonyms: Chroman-3-carboxylic acid

OR932278-1G OR932278-5G
APOSOR932278-1GEA 111 GBP
APOSOR932278-1G APOSOR932278-5G
3-Chromanecarboxylic acid 95%
3-Chromanecarboxylic acid
Formula: C₁₀H₁₀O₃
MW: 178,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03783554
CAS Number: 115822-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR