3-Chloro-5-methylbenzonitrile 97%

Supplier: Apollo Scientific

OR932275-10G OR932275-1G OR932275-5G
APOSOR932275-10GEA 814 GBP
APOSOR932275-10G APOSOR932275-1G APOSOR932275-5G
3-Chloro-5-methylbenzonitrile 97%
3-Chloro-5-methylbenzonitrile
Formula: C₈H₆ClN
MW: 151,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800704
CAS Number: 189161-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR