6-Methylthieno[2,3-d]pyrimidin-4(3H)-one 95%

Supplier: Apollo Scientific

Synonyms: 6-Methyl-3H-thieno[2,3-d]pyrimidin-4-one, 6-Methylthieno[2,3-d]pyrimidin-4-ol

OR932092-1G OR932092-5G
APOSOR932092-1GEA 39.3 GBP
APOSOR932092-1G APOSOR932092-5G
6-Methylthieno[2,3-d]pyrimidin-4(3H)-one 95%
6-Methylthieno[2,3-d]pyrimidin-4(3H)-one
Formula: C₇H₆N₂OS
MW: 166.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570294
CAS Number: 108831-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR