meso-Tetra(4-carboxyphenyl)porphine 97%

Supplier: Apollo Scientific

Synonyms: Tetrakis(4-carboxyphenyl)porphyrin, 4,4,4,4-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), TCPP, Tetrakis (4-carboxyphenyl) porphyrin

OR932015-1G OR932015-250MG OR932015-5G OR932015-25G
APOSOR932015-1GEA 59.1 GBP
APOSOR932015-1G APOSOR932015-250MG APOSOR932015-5G APOSOR932015-25G
meso-Tetra(4-carboxyphenyl)porphine 97%
meso-Tetra(4-carboxyphenyl)porphine
Formula: C₄₈H₃₀N₄O₈
MW: 790,79 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064860
CAS Number: 14609-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR