4-Bromo-N-methylaniline 98%
Supplier: Apollo Scientific
OR931639-25G
OR931639-5G
APOSOR931639-25GEA
212
GBP
APOSOR931639-25G
APOSOR931639-5G
4-Bromo-N-methylaniline 98%
4-Bromo-N-methylaniline
Formula:
C₇H₈BrN MW: 186,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03030391 CAS Number: 6911-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...