Vinyl butyrate 95%

Supplier: Apollo Scientific

Synonyms: Butyric Acid Vinyl Ester

OR930956-25G OR930956-500G
APOSOR930956-25GEA 0 GBP
APOSOR930956-25G APOSOR930956-500G
Vinyl butyrate 95%
Vinyl butyrate
Formula: C₆H₁₀O₂
MW: 114,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059410
CAS Number: 123-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR