Propyl cyanoacetate 95%

Supplier: Apollo Scientific

Synonyms: Propyl 2-cyanoacetate, Cyanoacetic Acid Propyl Ester

OR930774-25G OR930774-100G
APOSOR930774-25GEA 64.3 GBP
APOSOR930774-25G APOSOR930774-100G
Propyl cyanoacetate 95%
Propyl cyanoacetate
Formula: C₆H₉NO₂
MW: 127,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015784
CAS Number: 14447-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR