S-Propyl thioacetate 98%

Supplier: Apollo Scientific

Synonyms: S-Propyl ethanethioate

OR930740-100G OR930740-25G
APOSOR930740-100GEA 197 GBP
APOSOR930740-100G APOSOR930740-25G
S-Propyl thioacetate 98%
S-Propyl thioacetate
Formula: C₅H₁₀OS
MW: 118,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039937
CAS Number: 2307-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR