3',5'-Dimethylacetophenone 95%

Supplier: Apollo Scientific

Synonyms: 1-(3,5-Dimethylphenyl)ethanone, 1-(3,5-Dimethylphenyl)ethan-1-one

OR930709-10G OR930709-25G
APOSOR930709-10GEA 66.9 GBP
APOSOR930709-10G APOSOR930709-25G
3',5'-Dimethylacetophenone 95%
3',5'-Dimethylacetophenone
Formula: C₁₀H₁₂O
MW: 148,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075693
CAS Number: 5379-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR