5,6-Methyl-1,3-dihydro-2H-benzimidazol-2-one 95%

Supplier: Apollo Scientific

Synonyms: 5,6-Dimethyl-2-benzimidazolinone

OR930660-10G OR930660-1G OR930660-5G
APOSOR930660-10GEA 728 GBP
APOSOR930660-10G APOSOR930660-1G APOSOR930660-5G
5,6-Methyl-1,3-dihydro-2H-benzimidazol-2-one 95%
5,6-Methyl-1,3-dihydro-2H-benzimidazol-2-one
Formula: C₉H₁₀N₂O
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051725
CAS Number: 2033-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR