(3-Methyl-1H-pyrazol-1-yl)acetic acid 95%

Supplier: Apollo Scientific

OR930462-1G OR930462-250MG
APOSOR930462-1GEA 649 GBP
APOSOR930462-1G APOSOR930462-250MG
(3-Methyl-1H-pyrazol-1-yl)acetic acid 95%
(3-Methyl-1H-pyrazol-1-yl)acetic acid
Formula: C₆H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03419331
CAS Number: 180741-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR