3-Methyl-4-nitrobenzyl alcohol 95%

Supplier: Apollo Scientific

Synonyms: (3-Methyl-4-nitrophenyl)methanol, 5-(Hydroxymethyl)-2-nitrotoluene

OR930371-1G OR930371-5G
APOSOR930371-1GEA 59.1 GBP
APOSOR930371-1G APOSOR930371-5G
3-Methyl-4-nitrobenzyl alcohol 95%
3-Methyl-4-nitrobenzyl alcohol
Formula: C₈H₉NO₃
MW: 167,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007171
CAS Number: 80866-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR