(1-Indolyl)acetic acid 96%

Supplier: Apollo Scientific

Synonyms: Indole-1-acetic Acid, 2-(1H-Indol-1-yl)acetic acid

OR930357-10G OR930357-1G OR930357-5G
APOSOR930357-10GEA 68.3 GBP
APOSOR930357-10G APOSOR930357-1G APOSOR930357-5G
(1-Indolyl)acetic acid 96%
(1-Indolyl)acetic acid
Formula: C₁₀H₉NO₂
MW: 175,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047262
CAS Number: 24297-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR