Dimethyl-D-glutamate hydrochloride 97%

Supplier: Apollo Scientific

Synonyms: H-D-Glu(OMe)-OMe.HCl, H-D-Glu(OMe)-OMe·HCl, D-Glutamic Acid Dimethyl Ester Hydrochloride

OR930299-25G OR930299-5G
APOSOR930299-25GEA 175 GBP
APOSOR930299-25G APOSOR930299-5G
Dimethyl-D-glutamate hydrochloride 97%
Dimethyl-D-glutamate hydrochloride
Formula: C₇H₁₃NO₄
MW: 211,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235872
CAS Number: 27025-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR