Crotonoyl chloride mixture of isomers 96%

Supplier: Apollo Scientific

Synonyms: But-2-enoyl chloride, 2-Butenoyl chloride, Crotonoyl Chloride, Crotonic acid chloride

OR930056-100G OR930056-1KG OR930056-500G
APOSOR930056-100GEA 132 GBP
APOSOR930056-100G APOSOR930056-1KG APOSOR930056-500G
Crotonoyl chloride mixture of isomers 96%
Crotonoyl chloride
Formula: C₄H₅ClO
MW: 104,53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000734
CAS Number: 10487-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR