4-Phenoxybenzyl bromide 97%

Supplier: Apollo Scientific
Danger

Synonyms: 1-(Bromomethyl)-4-phenoxybenzene

OR9296-250MG OR9296-1G OR9296-5G
APOSOR9296-250MGEA 35.4 GBP
APOSOR9296-250MG APOSOR9296-1G APOSOR9296-5G
4-Phenoxybenzyl bromide 97%
4-Phenoxybenzyl bromide
Formula: C₁₃H₁₁BrO
Boiling Pt: 123 °C/0,5mmHg
Storage Temperature: Ambient
MDL Number: MFCD04038693
CAS Number: 36881-42-2
EINECS: 253-253-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR