Methyl-1-methylpiperidine-3-carboxylate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 1-methylpiperidine-3-carboxylate

OR929485-1G OR929485-5G
APOSOR929485-1GEA 46 GBP
APOSOR929485-1G APOSOR929485-5G
Methyl-1-methylpiperidine-3-carboxylate 95%
Methyl-1-methylpiperidine-3-carboxylate
Formula: C₈H₁₅NO₂
MW: 157,2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128793
CAS Number: 1690-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR